Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604969 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR1203 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR1203 - microRNA 1203 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SKAP1 - src kinase associated phosphoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001075099.1 | Intron | NP_001068567.1 | ||||
NM_003726.3 | Intron | NP_003717.3 | ||||
XM_005257755.3 | Intron | XP_005257812.1 | ||||
XM_017025257.1 | Intron | XP_016880746.1 | ||||
XM_017025258.1 | Intron | XP_016880747.1 | ||||
XM_017025259.1 | Intron | XP_016880748.1 | ||||
XM_017025260.1 | Intron | XP_016880749.1 |
Set Membership: |
HapMap |