Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATTTACGTATTCTAAATGGGAGG[C/T]GTAGTAGAAATGAACTTTAATATTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 614276 | |||||||||||||||||||||||
Literature Links: |
PLCL2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PLCL2 - phospholipase C like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144382.1 | 6962 | Intron | NP_001137854.1 | |||
NM_015184.5 | 6962 | Intron | NP_055999.2 | |||
XM_006713073.3 | 6962 | UTR 5 | XP_006713136.1 | |||
XM_017006022.1 | 6962 | Intron | XP_016861511.1 | |||
XM_017006023.1 | 6962 | Intron | XP_016861512.1 | |||
XM_017006024.1 | 6962 | Intron | XP_016861513.1 | |||
XM_017006025.1 | 6962 | UTR 5 | XP_016861514.1 |