Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGTTTGGGGCCTAGAAATGAACT[C/T]GGGAGGTTGTTTTGCTTGCTGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST3GAL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ST3GAL4 - ST3 beta-galactoside alpha-2,3-sialyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001254757.1 | Intron | NP_001241686.1 | ||||
NM_001254758.1 | Intron | NP_001241687.1 | ||||
NM_001254759.1 | Intron | NP_001241688.1 | ||||
NM_006278.2 | Intron | NP_006269.1 | ||||
XM_011542959.1 | Intron | XP_011541261.1 | ||||
XM_011542960.2 | Intron | XP_011541262.1 | ||||
XM_017018134.1 | Intron | XP_016873623.1 |
ST3GAL4-AS1 - ST3GAL4 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |