Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123810 MIM: 615257 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CREB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CREB1 - cAMP responsive element binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320793.1 | 5723 | Intron | NP_001307722.1 | |||
NM_004379.4 | 5723 | UTR 3 | NP_004370.1 | |||
NM_134442.4 | 5723 | UTR 3 | NP_604391.1 | |||
XM_011510645.1 | 5723 | UTR 3 | XP_011508947.1 | |||
XM_011510646.2 | 5723 | UTR 3 | XP_011508948.1 | |||
XM_011510647.2 | 5723 | UTR 3 | XP_011508949.1 | |||
XM_011510648.2 | 5723 | Intron | XP_011508950.1 | |||
XM_011510649.2 | 5723 | UTR 3 | XP_011508951.1 | |||
XM_011510650.2 | 5723 | UTR 3 | XP_011508952.1 | |||
XM_011510651.2 | 5723 | UTR 3 | XP_011508953.1 | |||
XM_017003399.1 | 5723 | Intron | XP_016858888.1 | |||
XM_017003400.1 | 5723 | UTR 3 | XP_016858889.1 | |||
XM_017003401.1 | 5723 | UTR 3 | XP_016858890.1 |