Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606622 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMARCAL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SMARCAL1 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127207.1 | 271 | Missense Mutation | ACT,GCT | T,A 43 | NP_001120679.1 | |
NM_014140.3 | 271 | Missense Mutation | ACT,GCT | T,A 43 | NP_054859.2 | |
XM_005246631.2 | 271 | Missense Mutation | ACT,GCT | T,A 43 | XP_005246688.1 | |
XM_005246632.1 | 271 | Missense Mutation | ACT,GCT | T,A 43 | XP_005246689.1 | |
XM_017004228.1 | 271 | UTR 5 | XP_016859717.1 |
Set Membership: |
HapMap |