Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 156490 MIM: 156491 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBTD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MBTD1 - mbt domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NME1 - NME/NM23 nucleoside diphosphate kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NME1-NME2 - NME1-NME2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018136.2 | 679 | UTR 3 | NP_001018146.1 |
NME2 - NME/NM23 nucleoside diphosphate kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018137.2 | 679 | UTR 3 | NP_001018147.1 | |||
NM_001018138.1 | 679 | Intron | NP_001018148.1 | |||
NM_001018139.2 | 679 | Intron | NP_001018149.1 | |||
NM_001198682.1 | 679 | UTR 3 | NP_001185611.1 | |||
NM_002512.3 | 679 | UTR 3 | NP_002503.1 |
Set Membership: |
HapMap |