Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGCTCCAATTATTAAATATTAC[C/T]GAGTACTTTTAGAAAATTGGGAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609314 MIM: 608094 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101930094 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101930094 - uncharacterized LOC101930094 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RSPH1 - radial spoke head 1 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC37A1 - solute carrier family 37 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320537.1 | Intron | NP_001307466.1 | ||||
NM_018964.3 | Intron | NP_061837.3 | ||||
XM_011529614.2 | Intron | XP_011527916.1 | ||||
XM_011529615.1 | Intron | XP_011527917.1 | ||||
XM_017028376.1 | Intron | XP_016883865.1 | ||||
XM_017028377.1 | Intron | XP_016883866.1 | ||||
XM_017028378.1 | Intron | XP_016883867.1 | ||||
XM_017028379.1 | Intron | XP_016883868.1 | ||||
XM_017028380.1 | Intron | XP_016883869.1 | ||||
XM_017028381.1 | Intron | XP_016883870.1 | ||||
XM_017028382.1 | Intron | XP_016883871.1 | ||||
XM_017028383.1 | Intron | XP_016883872.1 |
Set Membership: |
HapMap |