Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147150 MIM: 602060 | ||||||||||||||||||||
Literature Links: |
MX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MX1 - MX dynamin like GTPase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMPRSS2 - transmembrane protease, serine 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135099.1 | 1481 | Silent Mutation | AAC,AAG | N,K 486 | NP_001128571.1 | |
NM_005656.3 | 1481 | Silent Mutation | AAC,AAG | N,K 449 | NP_005647.3 | |
XM_005261043.2 | 1481 | Silent Mutation | AAC,AAG | N,K 409 | XP_005261100.1 | |
XM_011529731.2 | 1481 | Missense Mutation | AAC,AAG | N,K 449 | XP_011528033.1 | |
XM_011529732.2 | 1481 | Intron | XP_011528034.1 | |||
XM_011529733.1 | 1481 | Missense Mutation | AAC,AAG | N,K 341 | XP_011528035.1 |