Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615664 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TESPA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TESPA1 - thymocyte expressed, positive selection associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098815.2 | Intron | NP_001092285.1 | ||||
NM_001136030.2 | Intron | NP_001129502.1 | ||||
NM_001261844.1 | Intron | NP_001248773.1 | ||||
NM_014796.2 | Intron | NP_055611.1 | ||||
XM_005269247.2 | Intron | XP_005269304.1 | ||||
XM_006719715.3 | Intron | XP_006719778.1 | ||||
XM_011539035.2 | Intron | XP_011537337.1 | ||||
XM_011539036.1 | Intron | XP_011537338.1 | ||||
XM_011539037.2 | Intron | XP_011537339.1 | ||||
XM_017020262.1 | Intron | XP_016875751.1 | ||||
XM_017020263.1 | Intron | XP_016875752.1 |