Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606100 MIM: 611811 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGRE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADGRE2 - adhesion G protein-coupled receptor E2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271052.1 | Intron | NP_001257981.1 | ||||
NM_013447.3 | Intron | NP_038475.2 | ||||
XM_011527948.2 | Intron | XP_011526250.1 | ||||
XM_011527949.2 | Intron | XP_011526251.1 | ||||
XM_011527951.2 | Intron | XP_011526253.1 | ||||
XM_011527952.2 | Intron | XP_011526254.1 | ||||
XM_011527953.2 | Intron | XP_011526255.1 | ||||
XM_011527954.2 | Intron | XP_011526256.1 | ||||
XM_011527955.2 | Intron | XP_011526257.1 | ||||
XM_017026726.1 | Intron | XP_016882215.1 | ||||
XM_017026727.1 | Intron | XP_016882216.1 |
ZNF333 - zinc finger protein 333 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |