Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
60 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188410 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD1D PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CD1D - CD1d molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319145.1 | Intron | NP_001306074.1 | ||||
NM_001766.3 | Intron | NP_001757.1 | ||||
XM_005245583.1 | Intron | XP_005245640.1 | ||||
XM_006711621.1 | Intron | XP_006711684.1 | ||||
XM_011510125.1 | Intron | XP_011508427.1 | ||||
XM_011510127.1 | Intron | XP_011508429.1 |
Set Membership: |
HapMap |