Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
47 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134580 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
F13B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
F13B - coagulation factor XIII B chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001994.2 | 3133 | UTR 3 | NP_001985.2 | |||
XM_011509283.2 | 3133 | UTR 3 | XP_011507585.1 | |||
XM_011509284.2 | 3133 | UTR 3 | XP_011507586.1 | |||
XM_011509286.2 | 3133 | UTR 3 | XP_011507588.1 |
Set Membership: |
HapMap JSNP Validated |