Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCCCACATTGAGGGCAGCCCCCC[A/G]AGGTGAGATGGAGTGGCCTCCTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605492 | ||||||||||||||||||||
Literature Links: |
LOC105371692 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105371692 - uncharacterized LOC105371692 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRN2 - leucine rich repeat neuronal 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006338.2 | 3031 | UTR 3 | NP_006329.2 | |||
NM_201630.1 | 3031 | UTR 3 | NP_963924.1 | |||
XM_005244827.3 | 3031 | UTR 3 | XP_005244884.1 | |||
XM_011509066.2 | 3031 | UTR 3 | XP_011507368.1 | |||
XM_017000050.1 | 3031 | UTR 3 | XP_016855539.1 |