Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116810 MIM: 184420 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTSB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTSB - cathepsin B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317237.1 | 1939 | UTR 3 | NP_001304166.1 | |||
NM_001908.4 | 1939 | UTR 3 | NP_001899.1 | |||
NM_147780.3 | 1939 | UTR 3 | NP_680090.1 | |||
NM_147781.3 | 1939 | UTR 3 | NP_680091.1 | |||
NM_147782.3 | 1939 | UTR 3 | NP_680092.1 | |||
NM_147783.3 | 1939 | UTR 3 | NP_680093.1 | |||
XM_006716244.2 | 1939 | UTR 3 | XP_006716307.1 | |||
XM_006716245.2 | 1939 | UTR 3 | XP_006716308.1 | |||
XM_011543812.2 | 1939 | UTR 3 | XP_011542114.1 | |||
XM_017013097.1 | 1939 | UTR 3 | XP_016868586.1 | |||
XM_017013098.1 | 1939 | UTR 3 | XP_016868587.1 | |||
XM_017013099.1 | 1939 | UTR 3 | XP_016868588.1 | |||
XM_017013100.1 | 1939 | UTR 3 | XP_016868589.1 | |||
XM_017013101.1 | 1939 | UTR 3 | XP_016868590.1 |
FDFT1 - farnesyl-diphosphate farnesyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |