Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CNIH3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNIH3 - cornichon family AMPA receptor auxiliary protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322302.1 | Intron | NP_001309231.1 | ||||
NM_001322303.1 | Intron | NP_001309232.1 | ||||
NM_001322304.1 | Intron | NP_001309233.1 | ||||
NM_001322305.1 | Intron | NP_001309234.1 | ||||
NM_152495.1 | Intron | NP_689708.1 |
WDR26 - WD repeat domain 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |