Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612829 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB3C PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RAB3C - RAB3C, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317915.1 | Intron | NP_001304844.1 | ||||
NM_138453.3 | Intron | NP_612462.1 |
Set Membership: |
HapMap Validated |