Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAAGTCTCCGGCGCTGAGTGATG[C/T]CAACAAGGCCTCGGCCAGGGCCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600678 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MSH6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MSH6 - mutS homolog 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000179.2 | 211 | Missense Mutation | GCC,GTC | A,V 20 | NP_000170.1 | |
NM_001281492.1 | 211 | Missense Mutation | GCC,GTC | A,V 20 | NP_001268421.1 | |
NM_001281493.1 | 211 | UTR 5 | NP_001268422.1 | |||
NM_001281494.1 | 211 | Intron | NP_001268423.1 | |||
XM_011532798.1 | 211 | Intron | XP_011531100.1 | |||
XM_011532799.1 | 211 | Intron | XP_011531101.1 |