Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607758 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTNNBIP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CTNNBIP1 - catenin beta interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012329.1 | 3072 | UTR 3 | NP_001012329.1 | |||
NM_020248.2 | 3072 | UTR 3 | NP_064633.1 | |||
XM_017001853.1 | 3072 | UTR 3 | XP_016857342.1 | |||
XM_017001854.1 | 3072 | UTR 3 | XP_016857343.1 | |||
XM_017001855.1 | 3072 | UTR 3 | XP_016857344.1 |