Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616062 MIM: 614939 | ||||||||||||||||||||
Literature Links: |
ANKLE2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKLE2 - ankyrin repeat and LEM domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015114.2 | 2220 | Silent Mutation | GCC,GCT | A,A 937 | NP_055929.1 | |
XM_005266159.3 | 2220 | Silent Mutation | GCC,GCT | A,A 875 | XP_005266216.1 | |
XM_005266160.1 | 2220 | Silent Mutation | GCC,GCT | A,A 875 | XP_005266217.1 | |
XM_006719735.1 | 2220 | UTR 3 | XP_006719798.1 | |||
XM_011534787.2 | 2220 | Intron | XP_011533089.1 | |||
XM_011534788.1 | 2220 | Intron | XP_011533090.1 | |||
XM_017019076.1 | 2220 | Silent Mutation | GCC,GCT | A,A 500 | XP_016874565.1 |
PGAM5 - PGAM family member 5, mitochondrial serine/threonine protein phosphatase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |