Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605580 MIM: 600556 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL23A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
IL23A - interleukin 23 subunit alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAT2 - signal transducer and activator of transcription 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005419.3 | Intron | NP_005410.1 | ||||
NM_198332.1 | Intron | NP_938146.1 | ||||
XM_005269111.3 | Intron | XP_005269168.2 | ||||
XM_011538697.2 | Intron | XP_011536999.1 | ||||
XM_011538698.2 | Intron | XP_011537000.1 | ||||
XM_011538699.2 | Intron | XP_011537001.1 | ||||
XM_011538700.2 | Intron | XP_011537002.1 | ||||
XM_017019904.1 | Intron | XP_016875393.1 |
Set Membership: |
HapMap Validated |