Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACGCCTATTACAACCAGAAAACT[A/C]CAAGTATAACAGCGAGGATGGATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603180 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
XPOT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
XPOT - exportin for tRNA | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007235.4 | 489 | UTR 5 | NP_009166.2 | |||
XM_017018748.1 | 489 | UTR 5 | XP_016874237.1 |
Set Membership: |
HapMap |