Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605686 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CADM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CADM1 - cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098517.1 | 3331 | UTR 3 | NP_001091987.1 | |||
NM_001301043.1 | 3331 | UTR 3 | NP_001287972.1 | |||
NM_001301044.1 | 3331 | UTR 3 | NP_001287973.1 | |||
NM_001301045.1 | 3331 | UTR 3 | NP_001287974.1 | |||
NM_014333.3 | 3331 | UTR 3 | NP_055148.3 | |||
XM_005271494.2 | 3331 | UTR 3 | XP_005271551.1 | |||
XM_017017457.1 | 3331 | UTR 3 | XP_016872946.1 | |||
XM_017017458.1 | 3331 | UTR 3 | XP_016872947.1 | |||
XM_017017459.1 | 3331 | UTR 3 | XP_016872948.1 | |||
XM_017017460.1 | 3331 | UTR 3 | XP_016872949.1 | |||
XM_017017461.1 | 3331 | UTR 3 | XP_016872950.1 |
Set Membership: |
HapMap Validated |