Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
57 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613911 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF496 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF496 - zinc finger protein 496 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005273328.3 | 4840 | UTR 3 | XP_005273385.1 | |||
XM_005273330.3 | 4840 | UTR 3 | XP_005273387.1 | |||
XM_006711838.2 | 4840 | Intron | XP_006711901.1 | |||
XM_011544310.1 | 4840 | UTR 3 | XP_011542612.1 | |||
XM_011544312.1 | 4840 | UTR 3 | XP_011542614.1 | |||
XM_011544314.2 | 4840 | Intron | XP_011542616.1 | |||
XM_011544315.2 | 4840 | Intron | XP_011542617.1 | |||
XM_011544316.1 | 4840 | Intron | XP_011542618.1 | |||
XM_017002586.1 | 4840 | UTR 3 | XP_016858075.1 | |||
XM_017002587.1 | 4840 | UTR 3 | XP_016858076.1 | |||
XM_017002588.1 | 4840 | UTR 3 | XP_016858077.1 | |||
XM_017002589.1 | 4840 | UTR 3 | XP_016858078.1 | |||
XM_017002590.1 | 4840 | UTR 3 | XP_016858079.1 | |||
XM_017002591.1 | 4840 | UTR 3 | XP_016858080.1 | |||
XM_017002592.1 | 4840 | Intron | XP_016858081.1 | |||
XM_017002593.1 | 4840 | Intron | XP_016858082.1 | |||
XM_017002594.1 | 4840 | UTR 3 | XP_016858083.1 |
Set Membership: |
HapMap |