Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601643 | ||||||||||||||||||||
Literature Links: |
PPP2R5A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP2R5A - protein phosphatase 2 regulatory subunit B'alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM206 - transmembrane protein 206 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198862.1 | 1944 | UTR 3 | NP_001185791.1 | |||
NM_018252.2 | 1944 | UTR 3 | NP_060722.2 | |||
XM_005273177.3 | 1944 | UTR 3 | XP_005273234.1 | |||
XM_011509717.2 | 1944 | UTR 3 | XP_011508019.1 | |||
XM_011509718.2 | 1944 | UTR 3 | XP_011508020.1 | |||
XM_017001634.1 | 1944 | UTR 3 | XP_016857123.1 | |||
XM_017001635.1 | 1944 | UTR 3 | XP_016857124.1 | |||
XM_017001636.1 | 1944 | UTR 3 | XP_016857125.1 | |||
XM_017001637.1 | 1944 | UTR 3 | XP_016857126.1 |