Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176760 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRL PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRL - prolactin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000948.5 | 808 | Silent Mutation | GAA,GAG | E,E 190 | NP_000939.1 | |
NM_001163558.2 | 808 | Silent Mutation | GAA,GAG | E,E 190 | NP_001157030.1 | |
XM_011514753.2 | 808 | Silent Mutation | GAA,GAG | E,E 191 | XP_011513055.1 | |
XM_011514754.2 | 808 | Silent Mutation | GAA,GAG | E,E 191 | XP_011513056.1 |
Set Membership: |
HapMap |