Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608777 | ||||||||||||||||||||
Literature Links: |
POSTN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POSTN - periostin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135934.1 | 2855 | UTR 3 | NP_001129406.1 | |||
NM_001135935.1 | 2855 | UTR 3 | NP_001129407.1 | |||
NM_001135936.1 | 2855 | UTR 3 | NP_001129408.1 | |||
NM_001286665.1 | 2855 | UTR 3 | NP_001273594.1 | |||
NM_001286666.1 | 2855 | UTR 3 | NP_001273595.1 | |||
NM_001286667.1 | 2855 | UTR 3 | NP_001273596.1 | |||
NM_006475.2 | 2855 | UTR 3 | NP_006466.2 | |||
XM_005266231.2 | 2855 | UTR 3 | XP_005266288.1 | |||
XM_005266232.2 | 2855 | UTR 3 | XP_005266289.1 | |||
XM_017020355.1 | 2855 | Intron | XP_016875844.1 | |||
XM_017020356.1 | 2855 | Intron | XP_016875845.1 |