Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATTGATTAATTGCAAGCAGCTA[C/T]GTAGGGCACCCTGTGTTTAAATCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607019 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR548C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR548C - microRNA 548c | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR548Z - microRNA 548z | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASSF3 - Ras association domain family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178169.3 | Intron | NP_835463.1 | ||||
XM_006719345.3 | Intron | XP_006719408.1 | ||||
XM_011538195.2 | Intron | XP_011536497.1 | ||||
XM_017019182.1 | Intron | XP_016874671.1 |
Set Membership: |
HapMap |