Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGGAGGACGCAGGAGCTGCGGAG[A/C]CGGGCGCGAGGAGGAGGAGAGGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615856 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMTC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TMTC2 - transmembrane and tetratricopeptide repeat containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320321.1 | 452 | UTR 5 | NP_001307250.1 | |||
NM_001320322.1 | 452 | UTR 5 | NP_001307251.1 | |||
NM_152588.2 | 452 | UTR 5 | NP_689801.1 | |||
XM_017018884.1 | 452 | UTR 5 | XP_016874373.1 |
Set Membership: |
HapMap |