Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGACACAGTCGTCTGGGTCAGCA[T/C]GTAGTTCATTGGCGAGCATTAATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300446 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01402 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LINC01402 - long intergenic non-protein coding RNA 1402 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHOXF1 - Rhox homeobox family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139282.2 | 641 | Missense Mutation | CAT,CGT | H,R 172 | NP_644811.1 | |
XM_011531281.2 | 641 | Missense Mutation | CAT,CGT | H,R 200 | XP_011529583.1 |
RHOXF1-AS1 - RHOXF1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |