Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123811 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATF2 - activating transcription factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256090.1 | Intron | NP_001243019.1 | ||||
NM_001256091.1 | Intron | NP_001243020.1 | ||||
NM_001256092.1 | Intron | NP_001243021.1 | ||||
NM_001256093.1 | Intron | NP_001243022.1 | ||||
NM_001256094.1 | Intron | NP_001243023.1 | ||||
NM_001880.3 | Intron | NP_001871.2 |
Set Membership: |
HapMap |