Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAGCTGCTAGTCTGTCTCCAAA[A/G]AAAGTAAGTATGTTGAATAACATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613511 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102546294 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC102546294 - uncharacterized LOC102546294 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPINK7 - serine peptidase inhibitor, Kazal type 7 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032566.2 | 141 | Silent Mutation | AAA,AAG | K,K 28 | NP_115955.1 |
SPINK9 - serine peptidase inhibitor, Kazal type 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |