Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615124 | ||||||||||||||||||||
Literature Links: |
ANKRD13B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD13B - ankyrin repeat domain 13B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152345.4 | Intron | NP_689558.4 | ||||
XM_005257920.4 | Intron | XP_005257977.1 | ||||
XM_011524319.2 | Intron | XP_011522621.1 | ||||
XM_011524320.2 | Intron | XP_011522622.1 | ||||
XM_011524321.2 | Intron | XP_011522623.1 | ||||
XM_017024174.1 | Intron | XP_016879663.1 | ||||
XM_017024175.1 | Intron | XP_016879664.1 | ||||
XM_017024176.1 | Intron | XP_016879665.1 |