Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300463 MIM: 314375 MIM: 300249 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PQBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PQBP1 - polyglutamine binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032381.1 | Intron | NP_001027553.1 | ||||
NM_001032382.1 | Intron | NP_001027554.1 | ||||
NM_001032383.1 | Intron | NP_001027555.1 | ||||
NM_001032384.1 | Intron | NP_001027556.1 | ||||
NM_001167989.1 | Intron | NP_001161461.1 | ||||
NM_001167990.1 | Intron | NP_001161462.1 | ||||
NM_001167992.1 | Intron | NP_001161464.1 | ||||
NM_005710.2 | Intron | NP_005701.1 | ||||
NM_144495.2 | Intron | NP_652766.1 | ||||
XM_005272571.3 | Intron | XP_005272628.1 | ||||
XM_005272572.4 | Intron | XP_005272629.1 | ||||
XM_011543884.2 | Intron | XP_011542186.1 | ||||
XM_017029207.1 | Intron | XP_016884696.1 |
SLC35A2 - solute carrier family 35 member A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIMM17B - translocase of inner mitochondrial membrane 17 homolog B (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167947.2 | Intron | NP_001161419.1 | ||||
NM_005834.4 | Intron | NP_005825.1 |
Set Membership: |
HapMap Validated |