Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300019 MIM: 312420 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HCFC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HCFC1 - host cell factor C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005334.2 | 5535 | Silent Mutation | CTG,CTT | L,L 1514 | NP_005325.2 | |
XM_006724815.2 | 5535 | Silent Mutation | CTG,CTT | L,L 1558 | XP_006724878.1 | |
XM_006724816.2 | 5535 | Silent Mutation | CTG,CTT | L,L 1558 | XP_006724879.1 | |
XM_011531147.2 | 5535 | Silent Mutation | CTG,CTT | L,L 1514 | XP_011529449.1 | |
XM_011531148.2 | 5535 | Silent Mutation | CTG,CTT | L,L 1514 | XP_011529450.1 | |
XM_017029471.1 | 5535 | Silent Mutation | CTG,CTT | L,L 1492 | XP_016884960.1 | |
XM_017029472.1 | 5535 | Silent Mutation | CTG,CTT | L,L 1255 | XP_016884961.1 |
RENBP - renin binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |