Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108740 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP2A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATP2A2 - ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001681.3 | 644 | Silent Mutation | GAA,GAG | E,E 27 | NP_001672.1 | |
NM_170665.3 | 644 | Silent Mutation | GAA,GAG | E,E 27 | NP_733765.1 | |
XM_005253888.2 | 644 | Silent Mutation | GAA,GAG | E,E 27 | XP_005253945.1 | |
XM_011538402.2 | 644 | Silent Mutation | GAA,GAG | E,E 27 | XP_011536704.1 |
Set Membership: |
HapMap |