Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607926 MIM: 189904 | ||||||||||||||||||||
Literature Links: |
HCFC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HCFC2 - host cell factor C2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFYB - nuclear transcription factor Y subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006166.3 | 2659 | UTR 3 | NP_006157.1 | |||
XM_005268908.4 | 2659 | UTR 3 | XP_005268965.1 | |||
XM_005268909.4 | 2659 | UTR 3 | XP_005268966.1 | |||
XM_005268910.4 | 2659 | UTR 3 | XP_005268967.1 |