Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608426 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MKRN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MKRN2 - makorin ring finger protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271707.1 | 233 | Intron | NP_001258636.1 | |||
NM_014160.4 | 233 | Intron | NP_054879.3 |
MKRN2OS - MKRN2 opposite strand | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195279.1 | 233 | Intron | NP_001182208.1 | |||
XM_006712909.3 | 233 | UTR 5 | XP_006712972.1 | |||
XM_011533241.2 | 233 | Intron | XP_011531543.1 | |||
XM_011533242.2 | 233 | Intron | XP_011531544.1 | |||
XM_011533243.2 | 233 | Intron | XP_011531545.1 |
Set Membership: |
HapMap |