Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613004 | ||||||||||||||||||||
Literature Links: |
HTT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HTT - huntingtin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MSANTD1 - Myb/SANT DNA binding domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042690.1 | 4536 | Intron | NP_001036155.1 | |||
XM_011513465.2 | 4536 | Intron | XP_011511767.1 | |||
XM_011513466.2 | 4536 | Intron | XP_011511768.1 | |||
XM_011513467.2 | 4536 | UTR 5 | XP_011511769.1 | |||
XM_017008169.1 | 4536 | Intron | XP_016863658.1 |