Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCTCCTACATCAGGCATTTCAA[G/T]CTGTCAATATGTATTTATGATAGCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LACTB2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LACTB2 - lactamase beta 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LACTB2-AS1 - LACTB2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
XKR9 - XK related 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011720.1 | Intron | NP_001011720.1 | ||||
NM_001287258.1 | Intron | NP_001274187.1 | ||||
NM_001287259.1 | Intron | NP_001274188.1 | ||||
NM_001287260.1 | Intron | NP_001274189.1 | ||||
XM_006716447.3 | Intron | XP_006716510.1 | ||||
XM_011517524.2 | Intron | XP_011515826.1 | ||||
XM_011517525.2 | Intron | XP_011515827.1 | ||||
XM_011517526.2 | Intron | XP_011515828.1 | ||||
XM_011517527.2 | Intron | XP_011515829.1 | ||||
XM_017013404.1 | Intron | XP_016868893.1 | ||||
XM_017013405.1 | Intron | XP_016868894.1 |