Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142410 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf43 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C12orf43 - chromosome 12 open reading frame 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286191.1 | 1237 | UTR 3 | NP_001273120.1 | |||
NM_001286192.1 | 1237 | UTR 3 | NP_001273121.1 | |||
NM_001286195.1 | 1237 | UTR 3 | NP_001273124.1 | |||
NM_001286196.1 | 1237 | UTR 3 | NP_001273125.1 | |||
NM_001286197.1 | 1237 | UTR 3 | NP_001273126.1 | |||
NM_001286198.1 | 1237 | UTR 3 | NP_001273127.1 | |||
NM_022895.2 | 1237 | UTR 3 | NP_075046.1 | |||
XM_017019829.1 | 1237 | Intron | XP_016875318.1 |
HNF1A - HNF1 homeobox A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |