Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGATCCAGTTCATTCCACTCCCA[A/G]GAACCTGGGGCACAGAAGAGGTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600941 MIM: 612125 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLVRB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BLVRB - biliverdin reductase B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105372489 - uncharacterized LOC105372489 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SERTAD3 - SERTA domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013368.3 | 818 | Silent Mutation | TCC,TCT | S,S 180 | NP_037500.2 | |
NM_203344.2 | 818 | Silent Mutation | TCC,TCT | S,S 180 | NP_976219.1 | |
XM_005258832.3 | 818 | Silent Mutation | TCC,TCT | S,S 180 | XP_005258889.1 | |
XM_006723179.3 | 818 | Silent Mutation | TCC,TCT | S,S 180 | XP_006723242.1 |