Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 168468 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTH1R PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PTH1R - parathyroid hormone 1 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000316.2 | Intron | NP_000307.1 | ||||
NM_001184744.1 | Intron | NP_001171673.1 | ||||
XM_005265344.3 | Intron | XP_005265401.1 | ||||
XM_011533967.2 | Intron | XP_011532269.1 | ||||
XM_011533968.2 | Intron | XP_011532270.1 | ||||
XM_017006932.1 | Intron | XP_016862421.1 | ||||
XM_017006933.1 | Intron | XP_016862422.1 | ||||
XM_017006934.1 | Intron | XP_016862423.1 |
Set Membership: |
HapMap |