Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300576 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZDHHC15 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZDHHC15 - zinc finger DHHC-type containing 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146256.1 | Intron | NP_001139728.1 | ||||
NM_001146257.1 | Intron | NP_001139729.1 | ||||
NM_144969.2 | Intron | NP_659406.1 | ||||
XM_006724624.3 | Intron | XP_006724687.1 | ||||
XM_017029296.1 | Intron | XP_016884785.1 | ||||
XM_017029297.1 | Intron | XP_016884786.1 | ||||
XM_017029298.1 | Intron | XP_016884787.1 |