Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCTCTGGGAAGGGATGCTTGGC[C/T]TCCATGTTGATGCGCAGCCTCTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C21orf62 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C21orf62 - chromosome 21 open reading frame 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162495.2 | 687 | Silent Mutation | GAA,GAG | E,E 130 | NP_001155967.2 | |
NM_001162496.2 | 687 | Silent Mutation | GAA,GAG | E,E 130 | NP_001155968.2 | |
NM_019596.5 | 687 | Silent Mutation | GAA,GAG | E,E 130 | NP_062542.5 | |
XM_011529653.2 | 687 | Intron | XP_011527955.1 |
C21orf62-AS1 - C21orf62 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |