Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615165 MIM: 604972 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKIRIN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AKIRIN2 - akirin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018064.3 | Intron | NP_060534.1 |
ORC3 - origin recognition complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197259.1 | Intron | NP_001184188.1 | ||||
NM_012381.3 | Intron | NP_036513.2 | ||||
NM_181837.2 | Intron | NP_862820.1 | ||||
XM_005248704.1 | Intron | XP_005248761.1 | ||||
XM_005248705.1 | Intron | XP_005248762.1 | ||||
XM_005248706.1 | Intron | XP_005248763.1 | ||||
XM_011535651.1 | Intron | XP_011533953.1 | ||||
XM_011535652.2 | Intron | XP_011533954.1 | ||||
XM_017010632.1 | Intron | XP_016866121.1 | ||||
XM_017010633.1 | Intron | XP_016866122.1 | ||||
XM_017010634.1 | Intron | XP_016866123.1 | ||||
XM_017010635.1 | Intron | XP_016866124.1 | ||||
XM_017010636.1 | Intron | XP_016866125.1 | ||||
XM_017010637.1 | Intron | XP_016866126.1 | ||||
XM_017010638.1 | Intron | XP_016866127.1 | ||||
XM_017010639.1 | Intron | XP_016866128.1 | ||||
XM_017010640.1 | Intron | XP_016866129.1 |
Set Membership: |
HapMap |