Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 617010 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ULK4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ULK4 - unc-51 like kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322500.1 | Intron | NP_001309429.1 | ||||
NM_001322501.1 | Intron | NP_001309430.1 | ||||
NM_017886.3 | Intron | NP_060356.2 | ||||
XM_011533874.2 | Intron | XP_011532176.1 | ||||
XM_011533880.2 | Intron | XP_011532182.1 | ||||
XM_017006711.1 | Intron | XP_016862200.1 | ||||
XM_017006712.1 | Intron | XP_016862201.1 |
Set Membership: |
HapMap |