Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606765 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TPO PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TPO - thyroid peroxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000547.5 | 12 | UTR 5 | NP_000538.3 | |||
NM_001206744.1 | 12 | UTR 5 | NP_001193673.1 | |||
NM_001206745.1 | 12 | UTR 5 | NP_001193674.1 | |||
NM_175719.3 | 12 | UTR 5 | NP_783650.1 | |||
NM_175721.3 | 12 | Intron | NP_783652.1 | |||
NM_175722.3 | 12 | Intron | NP_783653.1 | |||
XM_011510379.2 | 12 | Intron | XP_011508681.1 | |||
XM_011510380.2 | 12 | Intron | XP_011508682.1 | |||
XM_011510381.2 | 12 | Intron | XP_011508683.1 |
Set Membership: |
HapMap |