Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602168 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VRK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
VRK1 - vaccinia related kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003384.2 | Intron | NP_003375.1 | ||||
XM_006720247.3 | Intron | XP_006720310.1 | ||||
XM_011537132.1 | Intron | XP_011535434.1 | ||||
XM_017021622.1 | Intron | XP_016877111.1 | ||||
XM_017021624.1 | Intron | XP_016877113.1 | ||||
XM_017021625.1 | Intron | XP_016877114.1 | ||||
XM_017021626.1 | Intron | XP_016877115.1 |
Set Membership: |
HapMap |