Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605242 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
USH1C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
USH1C - USH1 protein network component harmonin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297764.1 | Intron | NP_001284693.1 | ||||
NM_005709.3 | Intron | NP_005700.2 | ||||
NM_153676.3 | Intron | NP_710142.1 | ||||
XM_011519832.2 | Intron | XP_011518134.1 | ||||
XM_011519834.2 | Intron | XP_011518136.1 | ||||
XM_017017072.1 | Intron | XP_016872561.1 | ||||
XM_017017073.1 | Intron | XP_016872562.1 | ||||
XM_017017074.1 | Intron | XP_016872563.1 | ||||
XM_017017075.1 | Intron | XP_016872564.1 |
Set Membership: |
HapMap |