Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604480 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RINL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RINL - Ras and Rab interactor like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195833.1 | 1638 | Intron | NP_001182762.1 | |||
NM_198445.3 | 1638 | Intron | NP_940847.1 | |||
XM_006723015.2 | 1638 | Intron | XP_006723078.1 | |||
XM_006723016.2 | 1638 | Intron | XP_006723079.1 | |||
XM_011526454.2 | 1638 | Intron | XP_011524756.1 | |||
XM_011526455.2 | 1638 | Intron | XP_011524757.1 |
SIRT2 - sirtuin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193286.1 | 1638 | UTR 3 | NP_001180215.1 | |||
NM_012237.3 | 1638 | UTR 3 | NP_036369.2 | |||
NM_030593.2 | 1638 | UTR 3 | NP_085096.1 | |||
XM_006723111.1 | 1638 | UTR 3 | XP_006723174.1 | |||
XM_011526654.1 | 1638 | UTR 3 | XP_011524956.1 | |||
XM_011526655.1 | 1638 | UTR 3 | XP_011524957.1 | |||
XM_017026500.1 | 1638 | UTR 3 | XP_016881989.1 |
Set Membership: |
HapMap |